siRNA Details
Gene Symbol: WFDC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080614.1 | NP_542181.1 | 7 | 788 | ||
XM_011528553.2 | XP_011526855.1 | 7 | 798 | |||
GenBank mRNA | AF488306.1 | - | 7 | 788 | ||
AL591713.1 | - | 6 | 685 | |||
BC026014.1 | - | 5 | 557 | |||
BC102013.1 | - | 6 | 694 | |||
BC102014.1 | - | 4 | 430 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
