siRNA Details
Gene Symbol: CUL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015089.3 | NP_055904.1 | 7 | 1984 | ||
XM_011514422.1 | XP_011512724.1 | 7 | 1993 | |||
XM_011514423.2 | XP_011512725.1 | 8 | 2095 | |||
XM_011514424.1 | XP_011512726.1 | 7 | 1993 | |||
XM_011514425.1 | XP_011512727.1 | 5 | 1663 | |||
XM_011514426.1 | XP_011512728.1 | 7 | 1993 | |||
XM_011514427.1 | XP_011512729.1 | 3 | 612 | |||
XM_011514428.1 | XP_011512730.1 | 7 | 1993 | |||
XM_011514430.1 | XP_011512732.1 | 7 | 1993 | |||
XM_017010589.1 | XP_016866078.1 | 7 | 2704 | |||
GenBank mRNA | AJ318215.1 | - | 6 | 1909 | ||
AK125228.1 | - | 5 | 1655 | |||
AK129649.1 | - | 2 | 496 | |||
AY145132.1 | - | 6 | 1909 | |||
BC028159.1 | - | 7 | 1959 | |||
CR749511.1 | - | 7 | 1984 | |||
CR749841.1 | - | 7 | 1983 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
