siRNA Details
Gene Symbol: EXOC3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178516.3 | NP_848611.2 | 2, 3 | 298 | ||
GenBank mRNA | AK092858.1 | - | 2, 3 | 190 | ||
AK092906.1 | - | 3 | 245 | |||
AK292004.1 | - | 2, 3 | 207 | |||
AK300915.1 | - | 2, 3 | 266 | |||
BC034287.1 | - | 1, 2 | 480 | |||
DA945486.1 | - | 2, 3 | 298 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
