siRNA Details
Gene Symbol: CCDC74B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258307.1 | NP_001245236.1 | 8 | 1289 | ||
NM_207310.2 | NP_997193.1 | 8 | 1487 | |||
XM_005263840.2 | XP_005263897.1 | 8 | 1512 | |||
XM_005263842.2 | XP_005263899.1 | 8 | 1314 | |||
XM_006712833.2 | XP_006712896.1 | 8 | 1524 | |||
XM_006712834.3 | XP_006712897.1 | 8 | 1464 | |||
XM_006712835.2 | XP_006712898.1 | 8 | 1326 | |||
XM_006712837.2 | XP_006712900.1 | 8 | 1266 | |||
XM_011512142.2 | XP_011510444.1 | 6 | 1755 | |||
GenBank mRNA | AK057609.1 | - | 8 | 1411 | ||
AK297246.1 | - | 7 | 2399 | |||
AL133619.1 | - | 4 | 3323 | |||
BC067771.1 | - | 8 | 1234 | 4824929 | ||
CD359923.1 | - | 3 | 486 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
