siRNA Details
Gene Symbol: CCDC74B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258307.1 | NP_001245236.1 | 8 | 1140 | ||
NM_207310.2 | NP_997193.1 | 8 | 1338 | |||
XM_005263840.2 | XP_005263897.1 | 8 | 1363 | |||
XM_005263842.2 | XP_005263899.1 | 8 | 1165 | |||
XM_006712833.2 | XP_006712896.1 | 8 | 1375 | |||
XM_006712834.3 | XP_006712897.1 | 8 | 1315 | |||
XM_006712835.2 | XP_006712898.1 | 8 | 1177 | |||
XM_006712837.2 | XP_006712900.1 | 8 | 1117 | |||
XM_011512142.2 | XP_011510444.1 | 6 | 1606 | |||
GenBank mRNA | AK057609.1 | - | 8 | 1262 | ||
AK297246.1 | - | 7 | 2250 | |||
AL133619.1 | - | 4 | 3174 | |||
BC067771.1 | - | 8 | 1085 | 4824929 | ||
CD359923.1 | - | 3 | 337 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
