siRNA Details
Gene Symbol: LZTS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318099.1 | NP_001305028.1 | 3 | 606 | ||
NM_001318100.1 | NP_001305029.1 | 3 | 683 | |||
NM_032429.3 | NP_115805.1 | 3 | 713 | |||
XM_005270222.4 | XP_005270279.1 | 3 | 945 | |||
XM_017016780.1 | XP_016872269.1 | 2 | 3074 | |||
GenBank mRNA | AB058716.1 | - | 2 | 3565 | ||
AK097997.1 | - | 2 | 336 | |||
AK298208.1 | - | 3 | 571 | |||
AL834338.1 | - | 1 | 89 | |||
AY029201.1 | - | 2 | 427 | |||
BC006212.2 | - | 3 | 571 | 3161855 | ||
BC058938.1 | - | 3 | 646 | 3913470 | ||
BM544815.1 | - | 672 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
