siRNA Details
Gene Symbol: BBS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033604.1 | NP_001028776.1 | 20 | 2750 | ||
NM_001033605.1 | NP_001028777.1 | 20 | 2840 | |||
NM_014451.3 | NP_055266.2 | 19 | 2735 | |||
NM_198428.2 | NP_940820.1 | 21 | 2855 | |||
XM_005249700.3 | XP_005249757.1 | 21 | 2863 | |||
XM_011515264.2 | XP_011513566.1 | 21 | 2863 | |||
XM_011515265.2 | XP_011513567.1 | 21 | 2662 | |||
XM_011515266.2 | XP_011513568.1 | 20 | 2848 | |||
XM_011515267.2 | XP_011513569.1 | 20 | 2758 | |||
XM_011515268.1 | XP_011513570.1 | 21 | 2863 | |||
XM_011515269.2 | XP_011513571.1 | 21 | 2846 | |||
XM_017011990.1 | XP_016867479.1 | 20 | 2554 | |||
XM_017011991.1 | XP_016867480.1 | 21 | 2571 | |||
XM_017011992.1 | XP_016867481.1 | 21 | 2846 | |||
XM_017011993.1 | XP_016867482.1 | 20 | 2741 | |||
GenBank mRNA | AF095770.1 | - | 4 | 582 | ||
AF095771.1 | - | 18 | 2216 | |||
AK057660.1 | - | 12 | 1732 | |||
AK091361.1 | - | 8 | 1726 | |||
AK314627.1 | - | 21 | 2696 | |||
BC103831.2 | - | 21 | 2475 | 40001794 | ||
BX647385.1 | - | 22 | 2977 | |||
U85994.1 | - | 4 | 514 | |||
U85995.1 | - | 9 | 970 | |||
U87408.1 | - | 10 | 1041 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
