siRNA Details
Gene Symbol: BBS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033604.1 | NP_001028776.1 | 20 | 2843 | ||
NM_001033605.1 | NP_001028777.1 | 20 | 2933 | |||
NM_014451.3 | NP_055266.2 | 19 | 2828 | |||
NM_198428.2 | NP_940820.1 | 21 | 2948 | |||
XM_005249700.3 | XP_005249757.1 | 21 | 2956 | |||
XM_011515264.2 | XP_011513566.1 | 21 | 2956 | |||
XM_011515265.2 | XP_011513567.1 | 21 | 2755 | |||
XM_011515266.2 | XP_011513568.1 | 20 | 2941 | |||
XM_011515267.2 | XP_011513569.1 | 20 | 2851 | |||
XM_011515268.1 | XP_011513570.1 | 21 | 2956 | |||
XM_011515269.2 | XP_011513571.1 | 21 | 2939 | |||
XM_017011990.1 | XP_016867479.1 | 20 | 2647 | |||
XM_017011991.1 | XP_016867480.1 | 21 | 2664 | |||
XM_017011992.1 | XP_016867481.1 | 21 | 2939 | |||
XM_017011993.1 | XP_016867482.1 | 20 | 2834 | |||
GenBank mRNA | AF095770.1 | - | 4 | 675 | ||
AF095771.1 | - | 18 | 2309 | |||
AK057660.1 | - | 12 | 1825 | |||
AK091361.1 | - | 8 | 1819 | |||
AK314627.1 | - | 21 | 2789 | |||
BC103831.2 | - | 21 | 2568 | 40001794 | ||
BX647385.1 | - | 22 | 3070 | |||
U85994.1 | - | 4 | 607 | |||
U85995.1 | - | 9 | 1063 | |||
U87408.1 | - | 10 | 1134 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
