siRNA Details
Gene Symbol: NKAIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040214.2 | NP_001035304.1 | 6 | 950 | ||
NM_001300737.1 | NP_001287666.1 | 7 | 1014 | |||
NM_001300738.1 | NP_001287667.1 | 7 | 964 | |||
NM_001300740.1 | NP_001287669.1 | 6 | 588 | |||
NM_153355.4 | NP_699186.2 | 5 | 749 | |||
XM_011535501.2 | XP_011533803.1 | 6 | 838 | |||
XM_011535502.2 | XP_011533804.1 | 6 | 605 | |||
XM_011535503.2 | XP_011533805.1 | 5 | 875 | |||
XM_017010318.1 | XP_016865807.1 | 5 | 873 | |||
XM_017010319.1 | XP_016865808.1 | 5 | 637 | |||
GenBank mRNA | AK055326.1 | - | 6 | 583 | ||
AK309385.1 | - | 7 | 563 | |||
AK309407.1 | - | 3 | 325 | |||
AK309802.1 | - | 7 | 602 | |||
AK310763.1 | - | 5 | 424 | |||
AK315886.1 | - | 6 | 589 | |||
BC035062.2 | - | 7 | 716 | 5264304 | ||
DB174077.1 | - | 4 | 424 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
