siRNA Details
Gene Symbol: NKAIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040214.2 | NP_001035304.1 | 4 | 689 | ||
NM_001300737.1 | NP_001287666.1 | 5 | 753 | |||
NM_001300738.1 | NP_001287667.1 | 5 | 703 | |||
NM_001300740.1 | NP_001287669.1 | 4 | 327 | |||
XM_011535501.2 | XP_011533803.1 | 4 | 577 | |||
XM_011535502.2 | XP_011533804.1 | 4 | 344 | |||
XM_011535503.2 | XP_011533805.1 | 3 | 614 | |||
XM_017010318.1 | XP_016865807.1 | 3 | 612 | |||
GenBank mRNA | AB070452.1 | - | 4 | 498 | ||
AK055326.1 | - | 4 | 322 | |||
AK309385.1 | - | 5 | 302 | |||
AK309802.1 | - | 5 | 341 | |||
AK315886.1 | - | 4 | 328 | |||
BC035062.2 | - | 5 | 455 | 5264304 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
