siRNA Details
Gene Symbol: RPTOR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163034.1 | NP_001156506.1 | 30 | 5525 | ||
NM_020761.2 | NP_065812.1 | 34 | 5999 | |||
GenBank mRNA | AB037724.1 | - | 30 | 4681 | ||
AY090663.1 | - | 34 | 5556 | |||
BC014502.1 | - | 1 | 873 | |||
BC025180.1 | - | 5 | 1710 | |||
BC033258.1 | - | 10 | 2214 | |||
BC073898.1 | - | 4 | 1580 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
