siRNA Details
Gene Symbol: LRRC37BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124001.1 | - | 2 | 1146 | ||
AK124759.1 | - | 2 | 1553 | |||
AL713754.1 | - | 2 | 671 |
Gene Symbol: LOC107133515 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015341.2 | 9 | 1408 | |||
GenBank mRNA | AK075282.1 | - | 5 | 772 | ||
BC118647.1 | - | 5 | 891 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
