siRNA Details
Gene Symbol: XXYLT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308069.1 | NP_001294998.1 | 4 | 643 | ||
NM_152531.4 | NP_689744.3 | 4 | 1106 | |||
XM_005269286.4 | XP_005269343.1 | 4 | 3491 | |||
XM_011512448.2 | XP_011510750.1 | 5 | 918 | |||
XM_017005750.1 | XP_016861239.1 | 3 | 3879 | |||
GenBank mRNA | AK057046.1 | - | 5 | 851 | ||
AK075551.1 | - | 4 | 1074 | |||
AK092474.1 | - | 2 | 383 | |||
AK301133.1 | - | 4 | 405 | |||
BC019036.2 | - | 4 | 1015 | 4591786 | ||
BC039067.1 | - | 4 | 1088 | 5185557 | ||
BG721419.1 | - | 4 | 648 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
