siRNA Details
Gene Symbol: MSH5-SAPCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037846.1 | 26, 27 | 3470 | |||
GenBank mRNA | AJ245661.1 | - | 26, 27 | 3398 | ||
AL122094.1 | - | 11, 12 | 1623 |
Gene Symbol: SAPCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039651.1 | NP_001034740.1 | 2, 3 | 322 | ||
GenBank mRNA | BC137541.1 | - | 2, 3 | 287 | 9021541 | |
BC137542.1 | - | 2, 3 | 287 | 9021542 | ||
BU162048.1 | - | 2, 3 | 322 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
