siRNA Details
Gene Symbol: SAPCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039651.1 | NP_001034740.1 | 3 | 373 | ||
GenBank mRNA | BC137541.1 | - | 3 | 338 | 9021541 | |
BC137542.1 | - | 3 | 338 | 9021542 | ||
BU162048.1 | - | 3 | 373 |
Gene Symbol: MSH5-SAPCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037846.1 | 27 | 3521 | |||
GenBank mRNA | AJ245661.1 | - | 27 | 3449 | ||
AL122094.1 | - | 12 | 1674 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
