siRNA Details
Gene Symbol: LYRM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164841.2 | NP_001158313.1 | 4 | 1394 | ||
NM_020408.5 | NP_065141.3 | 3 | 1269 | |||
NR_104417.1 | 4 | 1616 | ||||
NR_104418.1 | 2 | 1148 | ||||
GenBank mRNA | AF258559.1 | - | 1 | 589 | ||
AK309458.1 | - | 3 | 1315 | |||
BC067762.1 | - | 2 | 940 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
