siRNA Details
Gene Symbol: SIGLEC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135163.1 | NP_001128635.1 | 7 | 1416 | ||
NM_052884.2 | NP_443116.2 | 7 | 1416 | |||
XM_005258476.3 | XP_005258533.2 | 6 | 1309 | |||
XM_005258477.3 | XP_005258534.2 | 7 | 1357 | |||
GenBank mRNA | AB072038.1 | - | 7 | 1335 | ||
AF337818.1 | - | 7 | 1416 | |||
AY358135.1 | - | 7 | 1294 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
