siRNA Details
Gene Symbol: DUOXA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207581.3 | NP_997464.2 | 5 | 1001 | ||
XM_017022180.1 | XP_016877669.1 | 5 | 1056 | |||
GenBank mRNA | AK303955.1 | - | 4 | 1362 | ||
BC137465.1 | - | 5 | 922 | 9021465 | ||
BX537581.1 | - | 5 | 1285 | |||
DQ489734.1 | - | 5 | 852 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
