siRNA Details
Gene Symbol: ZKSCAN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304506.1 | NP_001291435.1 | 5 | 617 | ||
NM_019110.4 | NP_061983.2 | 5 | 1282 | |||
XM_005249095.3 | XP_005249152.1 | 7 | 1326 | |||
XM_005249097.3 | XP_005249154.1 | 7 | 1022 | |||
XM_005249098.3 | XP_005249155.1 | 5 | 940 | |||
XM_005249100.4 | XP_005249157.2 | 5 | 673 | |||
XM_006715095.3 | XP_006715158.1 | 6 | 799 | |||
XM_011514588.2 | XP_011512890.1 | 5 | 1182 | |||
XM_011514589.2 | XP_011512891.1 | 5 | 738 | |||
XM_017010844.1 | XP_016866333.1 | 7 | 1977 | |||
GenBank mRNA | AK302017.1 | - | 5 | 598 | ||
AK315783.1 | - | 5 | 1252 | |||
AY781778.1 | - | 5 | 1252 | |||
BC014031.2 | - | 5 | 1210 | 4336904 | ||
KU178905.1 | - | 5 | 966 | |||
KU178906.1 | - | 4 | 855 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
