siRNA Details
Gene Symbol: TTC30A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152275.3 | NP_689488.3 | 1 | 2250 | ||
GenBank mRNA | AK057530.1 | - | 2 | 1887 | ||
AK125104.1 | - | 2 | 1954 | |||
AK292395.1 | - | 1 | 2234 | |||
AL049329.1 | - | 1 | 350 | |||
BC042848.1 | - | 1 | 2254 | 4831326 | ||
BC048006.1 | - | 1 | 2024 | 5299020 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
