siRNA Details
Gene Symbol: TTC30B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152517.2 | NP_689730.2 | 1 | 587 | ||
GenBank mRNA | BC033795.1 | - | 1 | 571 | 5175299 | |
BX647620.1 | - | 1 | 120 |
Gene Symbol: TTC30A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152275.3 | NP_689488.3 | 1 | 819 | ||
GenBank mRNA | AK057530.1 | - | 2 | 456 | ||
AK125104.1 | - | 2 | 523 | |||
AK292395.1 | - | 1 | 803 | |||
BC042848.1 | - | 1 | 823 | 4831326 | ||
BC048006.1 | - | 1 | 593 | 5299020 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
