siRNA Details
Gene Symbol: TMEM217 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286401.1 | NP_001273330.1 | 3 | 1895 | ||
NM_145316.3 | NP_660359.2 | 4 | 1462 | |||
NR_104442.1 | 3 | 1364 | ||||
NR_104443.1 | 2 | 1279 | ||||
XM_011514369.2 | XP_011512671.1 | 2 | 790 | |||
XM_011514370.2 | XP_011512672.1 | 2 | 780 | |||
GenBank mRNA | AK098666.1 | - | 4 | 1421 | ||
BC026012.1 | - | 3 | 1334 | 5269554 | ||
BC029657.1 | - | 2 | 760 | 5167418 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
