siRNA Details
Gene Symbol: SKIDA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207371.3 | NP_997254.3 | 4 | 3695 | ||
XM_005252450.3 | XP_005252507.1 | 5 | 2804 | |||
XM_011519480.2 | XP_011517782.1 | 5 | 2801 | |||
XM_017016225.1 | XP_016871714.1 | 5 | 3595 | |||
XM_017016226.1 | XP_016871715.1 | 5 | 3598 | |||
GenBank mRNA | AB234866.1 | - | 2 | 1296 | ||
AK127130.1 | - | 2 | 730 | |||
AK131456.1 | - | 5 | 3431 | |||
AK303207.1 | - | 3 | 1392 | |||
AL834446.1 | - | 1 | 413 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
