siRNA Details
Gene Symbol: ZNF524 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153219.3 | NP_694951.1 | 2 | 513 | ||
XM_011526487.2 | XP_011524789.1 | 2 | 754 | |||
GenBank mRNA | BC007396.2 | - | 1 | 396 | ||
BC014666.1 | - | 1 | 456 | 4864278 | ||
BC067748.1 | - | 2 | 484 | 5313025 | ||
BI756946.1 | - | 2 | 497 | |||
BM790568.1 | - | 2 | 488 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
