siRNA Details
Gene Symbol: STX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286210.1 | NP_001273139.1 | 6 | 1274 | ||
NM_005819.5 | NP_005810.1 | 8 | 1539 | |||
XM_017000007.1 | XP_016855496.1 | 7 | 1314 | |||
GenBank mRNA | AK056657.1 | - | 1 | 236 | ||
BC009944.2 | - | 8 | 1262 | 4122224 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
