siRNA Details
Gene Symbol: HNRNPD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003810.1 | NP_001003810.1 | 2 | 702 | ||
NM_002138.3 | NP_002129.2 | 3 | 759 | |||
NM_031369.2 | NP_112737.1 | 2 | 702 | |||
NM_031370.2 | NP_112738.1 | 3 | 759 | |||
GenBank mRNA | AF039575.1 | - | 3 | 417 | ||
AK057836.1 | - | 3 | 720 | |||
AK292707.1 | - | 2 | 697 | |||
AK300149.1 | - | 3 | 591 | |||
BC002401.1 | - | 3 | 505 | 2822230 | ||
BC023977.2 | - | 3 | 734 | 2961272 | ||
BC026015.2 | - | 3 | 734 | 2961272 | ||
D55671.1 | - | 2 | 178 | |||
D55672.1 | - | 3 | 726 | |||
D55673.1 | - | 2 | 169 | |||
D55674.1 | - | 3 | 705 | |||
M94630.1 | - | 3 | 399 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
