siRNA Details
Gene Symbol: ZNF347 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172674.1 | NP_001166145.1 | 5 | 1506 | ||
NM_001172675.1 | NP_001166146.1 | 5 | 1186 | |||
NM_032584.2 | NP_115973.2 | 5 | 1183 | |||
XM_005259335.4 | XP_005259392.1 | 5 | 1505 | |||
XM_005259336.4 | XP_005259393.1 | 5 | 1295 | |||
XM_017027384.1 | XP_016882873.1 | 5 | 1525 | |||
XM_017027385.1 | XP_016882874.1 | 5 | 1209 | |||
XM_017027386.1 | XP_016882875.1 | 4 | 1131 | |||
GenBank mRNA | AK095593.1 | - | 1 | 744 | ||
AK096623.1 | - | 5 | 1508 | |||
AK289994.1 | - | 5 | 1170 | |||
AK314603.1 | - | 5 | 1167 | |||
AL713691.1 | - | 1 | 165 | |||
AY029765.1 | - | 5 | 1145 | |||
BC136255.1 | - | 4 | 1135 | 9020242 | ||
BC143412.1 | - | 4 | 1138 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
