siRNA Details
Gene Symbol: FIP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134937.1 | NP_001128409.1 | 16 | 1651 | ||
NM_001134938.1 | NP_001128410.1 | 13 | 1447 | |||
NM_030917.3 | NP_112179.2 | 16 | 1669 | |||
XM_005265769.4 | XP_005265826.1 | 17 | 1763 | |||
XM_005265773.4 | XP_005265830.1 | 16 | 1696 | |||
XM_005265774.4 | XP_005265831.1 | 15 | 1608 | |||
XM_005265781.4 | XP_005265838.1 | 15 | 1580 | |||
XM_005265782.4 | XP_005265839.1 | 14 | 1499 | |||
XM_017008662.1 | XP_016864151.1 | 15 | 1641 | |||
XM_017008663.1 | XP_016864152.1 | 16 | 1597 | |||
XM_017008664.1 | XP_016864153.1 | 15 | 1652 | |||
XM_017008665.1 | XP_016864154.1 | 15 | 1568 | |||
XM_017008666.1 | XP_016864155.1 | 14 | 1585 | |||
XM_017008667.1 | XP_016864156.1 | 15 | 1551 | |||
XM_017008668.1 | XP_016864157.1 | 14 | 1597 | |||
XM_017008669.1 | XP_016864158.1 | 14 | 1497 | |||
GenBank mRNA | AK090938.1 | - | 5 | 454 | ||
AK123992.1 | - | 7 | 2167 | |||
AK295737.1 | - | 16 | 1666 | |||
AL136910.1 | - | 13 | 1428 | |||
AY366510.1 | - | 16 | 1626 | |||
BC011543.1 | - | 7 | 687 | |||
BC024016.1 | - | 14 | 2711 | 4826276 | ||
BC026724.1 | - | 5 | 491 | |||
BC052959.1 | - | 14 | 2706 | 4838737 | ||
BC110383.1 | - | 16 | 1662 | 5165430 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
