siRNA Details
Gene Symbol: C19orf60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100418.1 | NP_001093888.1 | 5 | 855 | ||
NM_001100419.1 | NP_001093889.1 | 5 | 789 | |||
XM_005259977.1 | XP_005260034.1 | 3 | 776 | |||
GenBank mRNA | AK000857.1 | - | 5 | 743 | ||
AK054670.1 | - | 4 | 1947 | |||
AK091925.1 | - | 3 | 2251 | |||
AK094710.1 | - | 1 | 2442 | |||
BC012078.1 | - | 5 | 744 | 4661993 | ||
BI836487.1 | - | 5 | 666 | |||
KP143545.1 | - | 5 | 649 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
