siRNA Details
Gene Symbol: SSU72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014188.2 | NP_054907.1 | 1, 2 | 402 | ||
GenBank mRNA | AF161530.1 | - | 1, 2 | 142 | ||
AF277178.1 | - | 1, 2 | 81 | |||
AJ276409.1 | - | 1, 2 | 190 | |||
AK001809.1 | - | 1, 2 | 150 | |||
AK023110.1 | - | 1, 2 | 104 | |||
AK291324.1 | - | 1, 2 | 152 | |||
AK297505.1 | - | 1, 2 | 147 | |||
AK310067.1 | - | 1, 2 | 151 | |||
BC008070.2 | - | 1, 2 | 350 | 2961422 | ||
BC018909.2 | - | 1, 2 | 386 | |||
CF594026.1 | - | 432 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
