siRNA Details
Gene Symbol: SSU72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014188.2 | NP_054907.1 | 1 | 374 | ||
GenBank mRNA | AF161530.1 | - | 1 | 114 | ||
AF277178.1 | - | 1 | 53 | |||
AJ276409.1 | - | 1 | 162 | |||
AK001809.1 | - | 1 | 122 | |||
AK023110.1 | - | 1 | 76 | |||
AK291324.1 | - | 1 | 124 | |||
AK297505.1 | - | 1 | 119 | |||
AK310067.1 | - | 1 | 123 | |||
BC008070.2 | - | 1 | 322 | 2961422 | ||
BC018909.2 | - | 1 | 358 | |||
CF594026.1 | - | 404 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
