siRNA Details
Gene Symbol: SSU72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014188.2 | NP_054907.1 | 2 | 475 | ||
GenBank mRNA | AF161530.1 | - | 2 | 215 | ||
AF277178.1 | - | 2 | 154 | |||
AJ276409.1 | - | 2 | 263 | |||
AK001809.1 | - | 2 | 223 | |||
AK023110.1 | - | 2 | 177 | |||
AK291324.1 | - | 2 | 225 | |||
AK297505.1 | - | 2 | 220 | |||
AK310067.1 | - | 2 | 224 | |||
BC008070.2 | - | 2 | 423 | 2961422 | ||
BC018909.2 | - | 2 | 459 | |||
CF594026.1 | - | 505 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
