siRNA Details
Gene Symbol: ANKRD12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083625.2 | NP_001077094.1 | 8 | 2618 | ||
NM_001204056.1 | NP_001190985.1 | 8 | 2716 | |||
NM_015208.4 | NP_056023.3 | 9 | 2687 | |||
XM_005258092.3 | XP_005258149.1 | 9 | 2820 | |||
XM_005258093.3 | XP_005258150.1 | 9 | 2804 | |||
XM_005258095.2 | XP_005258152.1 | 9 | 2653 | |||
XM_005258096.2 | XP_005258153.1 | 8 | 2588 | |||
XM_011525638.2 | XP_011523940.1 | 9 | 2659 | |||
XM_011525639.2 | XP_011523941.1 | 10 | 2850 | |||
XM_017025661.1 | XP_016881150.1 | 9 | 2782 | |||
XM_017025662.1 | XP_016881151.1 | 9 | 2827 | |||
XM_017025663.1 | XP_016881152.1 | 8 | 2703 | |||
XM_017025664.1 | XP_016881153.1 | 8 | 2697 | |||
XM_017025665.1 | XP_016881154.1 | 8 | 2731 | |||
XM_017025666.1 | XP_016881155.1 | 8 | 2670 | |||
XM_017025667.1 | XP_016881156.1 | 10 | 2681 | |||
XM_017025668.1 | XP_016881157.1 | 10 | 2675 | |||
XM_017025669.1 | XP_016881158.1 | 3 | 1777 | |||
XM_017025670.1 | XP_016881159.1 | 5 | 2244 | |||
GenBank mRNA | AB075493.1 | - | 4 | 1830 | ||
AF317425.1 | - | 8 | 2423 | |||
AK000060.1 | - | 4 | 1815 | |||
AL834204.1 | - | 8 | 2546 | |||
AY373757.1 | - | 9 | 2637 | |||
AY533564.2 | - | 8 | 2716 | |||
X80821.1 | - | 2 | 435 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
