siRNA Details
Gene Symbol: TLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006289.3 | NP_006280.3 | 57 | 8023 | ||
GenBank mRNA | AB028950.2 | - | 57 | 7998 | ||
AF078828.1 | - | 57 | 7795 | |||
AF113217.1 | - | 2 | 245 | |||
AF177198.1 | - | 57 | 7759 | |||
BC042923.1 | - | 57 | 7809 | 5576203 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
