siRNA Details
Gene Symbol: LHX8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001933.1 | NP_001001933.1 | 5, 6 | 1053 | ||
NM_001256114.1 | NP_001243043.1 | 4, 5 | 444 | |||
XM_017001316.1 | XP_016856805.1 | 4, 5 | 933 | |||
XM_017001317.1 | XP_016856806.1 | 3, 4 | 453 | |||
GenBank mRNA | BC040321.1 | - | 5, 6 | 1053 | 4839343 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
