siRNA Details
Gene Symbol: PPHLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143788.1 | NP_001137260.1 | 3, 4 | 348 | ||
NM_016488.6 | NP_057572.5 | 3, 4 | 348 | |||
NM_201439.1 | NP_958847.1 | 3, 4 | 348 | |||
NM_201515.1 | NP_958923.1 | 4, 5 | 391 | |||
XM_011538459.2 | XP_011536761.1 | 4, 5 | 369 | |||
XM_011538465.2 | XP_011536767.1 | 3, 4 | 326 | |||
XM_017019434.1 | XP_016874923.1 | 4, 5 | 369 | |||
XM_017019435.1 | XP_016874924.1 | 4, 5 | 368 | |||
XM_017019436.1 | XP_016874925.1 | 4, 5 | 369 | |||
XM_017019437.1 | XP_016874926.1 | 3, 4 | 326 | |||
XM_017019438.1 | XP_016874927.1 | 3, 4 | 326 | |||
XM_017019439.1 | XP_016874928.1 | 4, 5 | 369 | |||
XM_017019440.1 | XP_016874929.1 | 4, 5 | 369 | |||
XM_017019441.1 | XP_016874930.1 | 3, 4 | 326 | |||
XM_017019442.1 | XP_016874931.1 | 3, 4 | 326 | |||
XM_017019443.1 | XP_016874932.1 | 4, 5 | 369 | |||
XM_017019452.1 | XP_016874941.1 | 4, 5 | 369 | |||
GenBank mRNA | AK055690.1 | - | 3, 4 | 336 | ||
AK129910.1 | - | 4, 5 | 391 | |||
AK308022.1 | - | 3, 4 | 322 | |||
AY039238.1 | - | 3, 4 | 330 | |||
AY157850.1 | - | 4, 5 | 387 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
