siRNA Details
Gene Symbol: NKX2-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079668.2 | NP_001073136.1 | 2 | 425 | ||
NM_003317.3 | NP_003308.1 | 1 | 580 | |||
GenBank mRNA | BC006221.2 | - | 2 | 359 | 3941576 | |
BE796718.1 | - | 2 | 340 | |||
BT009773.1 | - | 2 | 329 | |||
D50740.1 | - | 3 | 483 | |||
D50741.1 | - | 1 | 300 | |||
U19756.1 | - | 1 | 239 | |||
U33749.1 | - | 1 | 580 | |||
U43203.1 | - | 2 | 411 | |||
X82850.1 | - | 1 | 253 |

More Information
Other Species Targeted

Gene Ontology Categories:
