siRNA Details
Gene Symbol: NKX2-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079668.2 | NP_001073136.1 | 3 | 1167 | ||
NM_003317.3 | NP_003308.1 | 2 | 1322 | |||
GenBank mRNA | BC006221.2 | - | 3 | 1101 | 3941576 | |
BT009773.1 | - | 3 | 1071 | |||
D50740.1 | - | 4 | 1225 | |||
D50741.1 | - | 1 | 1994 | |||
U19756.1 | - | 2 | 981 | |||
U33749.1 | - | 2 | 1322 | |||
U43203.1 | - | 3 | 1153 | |||
X82850.1 | - | 2 | 995 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
