siRNA Details
Gene Symbol: DCN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001920.4 | NP_001911.1 | 2 | 525 | ||
NM_133503.3 | NP_598010.1 | 2 | 312 | |||
NM_133504.3 | NP_598011.1 | 1 | 116 | |||
NM_133505.3 | NP_598012.1 | 1 | 116 | |||
NM_133506.3 | NP_598013.1 | 1 | 116 | |||
NM_133507.3 | NP_598014.1 | 1 | 116 | |||
XM_005268693.1 | XP_005268750.1 | 2 | 278 | |||
XM_006719270.1 | XP_006719333.1 | 2 | 101 | |||
XM_017018917.1 | XP_016874406.1 | 2 | 252 | |||
GenBank mRNA | AF138300.1 | - | 1 | 22 | ||
AF138301.1 | - | 1 | 22 | |||
AF138302.1 | - | 1 | 22 | |||
AF138303.1 | - | 1 | 22 | |||
AF138304.1 | - | 1 | 22 | |||
AK291309.1 | - | 2 | 183 | |||
AL355731.1 | - | 2 | 203 | |||
BC005322.1 | - | 1 | 63 | 3934022 | ||
BG536640.1 | - | 278 | ||||
BG538290.1 | - | 2 | 275 | |||
BT019800.1 | - | 1 | 22 | |||
BX571756.1 | - | 2 | 239 | |||
CD689331.1 | - | 2 | 536 | |||
CR407676.1 | - | 1 | 22 | |||
CR541946.1 | - | 1 | 22 | |||
DA424244.1 | - | 1 | 89 | |||
DA424310.1 | - | 2 | 312 | |||
DA663155.1 | - | 2 | 527 | |||
KU178017.1 | - | 1 | 22 | |||
KU178018.1 | - | 1 | 22 | |||
KU178019.1 | - | 1 | 22 | |||
KU178020.1 | - | 1 | 22 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
