siRNA Details
Gene Symbol: BCLAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077440.1 | NP_001070908.1 | 5 | 1847 | ||
NM_001077441.1 | NP_001070909.1 | 5 | 1334 | |||
NM_001301038.1 | NP_001287967.1 | 5 | 1847 | |||
NM_014739.2 | NP_055554.1 | 5 | 1853 | |||
XM_005267237.3 | XP_005267294.1 | 5 | 1789 | |||
XM_017011529.1 | XP_016867018.1 | 5 | 1321 | |||
XM_017011530.1 | XP_016867019.1 | 5 | 2128 | |||
XM_017011531.1 | XP_016867020.1 | 5 | 1264 | |||
XM_017011532.1 | XP_016867021.1 | 5 | 1789 | |||
GenBank mRNA | AF249273.1 | - | 3 | 1594 | ||
BC016682.1 | - | 5 | 1792 | |||
BC047687.1 | - | 4 | 1160 | |||
BC047887.1 | - | 5 | 1822 | |||
BC056894.1 | - | 5 | 1834 | |||
BC063846.1 | - | 5 | 1760 | |||
BC078160.1 | - | 5 | 1759 | |||
BC132780.1 | - | 5 | 1797 | 40146802 | ||
BC144281.1 | - | 5 | 1791 | 9052801 | ||
BC144299.1 | - | 5 | 1797 | |||
BX648878.1 | - | 1 | 594 | |||
BX649160.1 | - | 5 | 1265 | |||
D79986.1 | - | 5 | 1853 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
