siRNA Details
Gene Symbol: BCLAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077440.1 | NP_001070908.1 | 10 | 2540 | ||
NM_001077441.1 | NP_001070909.1 | 10 | 2027 | |||
NM_001301038.1 | NP_001287967.1 | 10 | 2540 | |||
NM_014739.2 | NP_055554.1 | 10 | 2546 | |||
XM_005267237.3 | XP_005267294.1 | 10 | 2482 | |||
XM_017011529.1 | XP_016867018.1 | 10 | 2014 | |||
XM_017011530.1 | XP_016867019.1 | 10 | 2821 | |||
XM_017011531.1 | XP_016867020.1 | 10 | 1957 | |||
XM_017011532.1 | XP_016867021.1 | 11 | 2709 | |||
GenBank mRNA | AF249273.1 | - | 8 | 2287 | ||
AK303559.1 | - | 1 | 406 | |||
AK308870.1 | - | 3 | 537 | |||
AK309259.1 | - | 3 | 760 | |||
AK315884.1 | - | 2 | 144 | |||
BC132780.1 | - | 10 | 2490 | 40146802 | ||
BC144281.1 | - | 10 | 2484 | 9052801 | ||
BC144299.1 | - | 11 | 2654 | |||
BX648431.1 | - | 2 | 208 | |||
BX648878.1 | - | 6 | 1287 | |||
BX649160.1 | - | 10 | 1958 | |||
D79986.1 | - | 10 | 2546 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
