siRNA Details
Gene Symbol: NAIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004536.2 | NP_004527.2 | 17 | 5557 | ||
NM_022892.1 | NP_075043.1 | 15 | 4886 | |||
XM_011543413.2 | XP_011541715.1 | 7 | 3695 | |||
XM_017009490.1 | XP_016864979.1 | 7 | 4242 | |||
GenBank mRNA | AB209123.1 | - | 6 | 4943 | ||
U19251.1 | - | 17 | 5130 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
