siRNA Details
Gene Symbol: RBM41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171080.1 | NP_001164551.1 | 4 | 407 | ||
NM_001324242.1 | NP_001311171.1 | 4 | 407 | |||
NM_001324243.1 | NP_001311172.1 | 4 | 500 | |||
NM_001324244.1 | NP_001311173.1 | 4 | 407 | |||
NM_018301.4 | NP_060771.3 | 4 | 407 | |||
NR_136728.1 | 4 | 407 | ||||
NR_136729.1 | 5 | 501 | ||||
NR_136730.1 | 4 | 407 | ||||
NR_136731.1 | 4 | 500 | ||||
NR_136732.1 | 4 | 500 | ||||
XM_011530980.2 | XP_011529282.1 | 4 | 499 | |||
XM_011530981.2 | XP_011529283.1 | 4 | 499 | |||
GenBank mRNA | AK001878.1 | - | 4 | 380 | ||
AK023732.1 | - | 4 | 403 | |||
BC006986.1 | - | 4 | 384 | 3995903 | ||
DA255574.1 | - | 4 | 407 | |||
DB132398.1 | - | 4 | 407 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
