siRNA Details
Gene Symbol: RPGRIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020366.3 | NP_065099.3 | 20, 21 | 3336 | ||
XM_005267879.2 | XP_005267936.1 | 11, 12 | 2331 | |||
XM_005267880.2 | XP_005267937.1 | 11, 12 | 2298 | |||
XM_005267881.3 | XP_005267938.1 | 9, 10 | 1826 | |||
XM_011536978.1 | XP_011535280.1 | 11, 12 | 2328 | |||
XM_011536979.1 | XP_011535281.1 | 12, 13 | 2115 | |||
XM_011536980.1 | XP_011535282.1 | 11, 12 | 1986 | |||
XM_011536981.1 | XP_011535283.1 | 9, 10 | 1836 | |||
XM_011536982.1 | XP_011535284.1 | 9, 10 | 1491 | |||
XM_017021473.1 | XP_016876962.1 | 9, 10 | 1833 | |||
GenBank mRNA | AF227257.1 | - | 9, 10 | 1764 | ||
AF260257.1 | - | 11, 12 | 2288 | |||
AF265666.1 | - | 9, 10 | 1763 | |||
AF265667.1 | - | 9, 10 | 1529 | |||
AJ417067.1 | - | 20, 21 | 3336 | |||
AK301780.1 | - | 8, 9 | 1394 | |||
BC039089.1 | - | 8, 9 | 1379 | |||
BX571740.1 | - | 7, 8 | 2268 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
