siRNA Details
Gene Symbol: RPGRIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020366.3 | NP_065099.3 | 21 | 3519 | ||
XM_005267879.2 | XP_005267936.1 | 12 | 2514 | |||
XM_005267880.2 | XP_005267937.1 | 12 | 2481 | |||
XM_005267881.3 | XP_005267938.1 | 10 | 2009 | |||
XM_011536978.1 | XP_011535280.1 | 12 | 2511 | |||
XM_011536979.1 | XP_011535281.1 | 13 | 2298 | |||
XM_011536980.1 | XP_011535282.1 | 12 | 2169 | |||
XM_011536981.1 | XP_011535283.1 | 10 | 2019 | |||
XM_011536982.1 | XP_011535284.1 | 10 | 1674 | |||
XM_017021473.1 | XP_016876962.1 | 10 | 2016 | |||
GenBank mRNA | AF227257.1 | - | 10 | 1947 | ||
AF260257.1 | - | 12 | 2471 | |||
AF265666.1 | - | 10 | 1946 | |||
AF265667.1 | - | 10 | 1712 | |||
AJ417067.1 | - | 21 | 3519 | |||
AK301780.1 | - | 9 | 1577 | |||
BC039089.1 | - | 9 | 1562 | |||
BX571740.1 | - | 8 | 2451 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
