siRNA Details
Gene Symbol: RPGRIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020366.3 | NP_065099.3 | 14 | 2082 | ||
XM_005267879.2 | XP_005267936.1 | 5 | 1074 | |||
XM_005267880.2 | XP_005267937.1 | 5 | 1041 | |||
XM_005267881.3 | XP_005267938.1 | 3 | 569 | |||
XM_011536978.1 | XP_011535280.1 | 5 | 1074 | |||
XM_011536981.1 | XP_011535283.1 | 5 | 1074 | |||
XM_017021473.1 | XP_016876962.1 | 5 | 1074 | |||
GenBank mRNA | AF227257.1 | - | 3 | 510 | ||
AF260257.1 | - | 5 | 1034 | |||
AF265666.1 | - | 3 | 509 | |||
AF265667.1 | - | 3 | 275 | |||
AJ417067.1 | - | 14 | 2082 | |||
BX571740.1 | - | 1 | 1014 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
