siRNA Details
Gene Symbol: CRYBB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000496.2 | NP_000487.1 | 3 | 141 | ||
XM_006724141.3 | XP_006724204.1 | 3 | 1300 | |||
XM_011529900.2 | XP_011528202.1 | 4 | 1665 | |||
GenBank mRNA | BC069535.1 | - | 2 | 105 | 7262188 | |
CR456426.1 | - | 2 | 103 | |||
KR052782.1 | - | 3 | 135 | |||
KR052783.1 | - | 3 | 135 | |||
KR052784.1 | - | 3 | 137 | |||
KR052788.1 | - | 2 | 93 | |||
KR052789.1 | - | 2 | 84 | |||
KR052790.1 | - | 2 | 126 | |||
KR052791.1 | - | 1 | 41 | |||
L10035.1 | - | 2 | 105 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
