siRNA Details
Gene Symbol: CRYBB2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033733.1 | 3, 4 | 447 | |||
NR_033734.1 | 2, 3 | 328 | ||||
GenBank mRNA | BC037884.1 | - | 2, 3 | 315 | ||
BC047380.1 | - | 3, 4 | 450 |
Gene Symbol: CRYBB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000496.2 | NP_000487.1 | 4, 5 | 355 | ||
XM_006724141.3 | XP_006724204.1 | 4, 5 | 1514 | |||
XM_011529900.2 | XP_011528202.1 | 5, 6 | 1879 | |||
GenBank mRNA | BC069535.1 | - | 3, 4 | 319 | 7262188 | |
CR456426.1 | - | 3, 4 | 317 | |||
KR052782.1 | - | 4, 5 | 349 | |||
KR052783.1 | - | 4, 5 | 349 | |||
KR052784.1 | - | 4, 5 | 351 | |||
KR052788.1 | - | 3, 4 | 307 | |||
KR052789.1 | - | 3, 4 | 298 | |||
KR052790.1 | - | 3, 4 | 340 | |||
KR052791.1 | - | 2, 3 | 255 | |||
L10035.1 | - | 3, 4 | 319 |

More Information
Other Species Targeted

Gene Ontology Categories:
