siRNA Details
Gene Symbol: CRYBB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000496.2 | NP_000487.1 | 5 | 483 | ||
XM_006724141.3 | XP_006724204.1 | 5 | 1642 | |||
XM_011529900.2 | XP_011528202.1 | 6 | 2007 | |||
GenBank mRNA | BC069535.1 | - | 4 | 447 | 7262188 | |
CR456426.1 | - | 4 | 445 | |||
KR052782.1 | - | 5 | 477 | |||
KR052783.1 | - | 5 | 477 | |||
KR052784.1 | - | 5 | 479 | |||
KR052788.1 | - | 4 | 435 | |||
KR052789.1 | - | 4 | 426 | |||
KR052790.1 | - | 4 | 468 | |||
KR052791.1 | - | 3 | 383 | |||
L10035.1 | - | 4 | 447 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
