siRNA Details
Gene Symbol: RIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024832.4 | NP_079108.3 | 2 | 384 | ||
XM_011537165.2 | XP_011535467.1 | 2 | 392 | |||
XM_017021651.1 | XP_016877140.1 | 2 | 266 | |||
XM_017021652.1 | XP_016877141.1 | 2 | 392 | |||
XM_017021653.1 | XP_016877142.1 | 2 | 392 | |||
XM_017021654.1 | XP_016877143.1 | 2 | 392 | |||
GenBank mRNA | AB081753.1 | - | 2 | 232 | ||
AK021762.1 | - | 2 | 384 | |||
BC070062.1 | - | 2 | 372 | 30332851 | ||
BX248258.1 | - | 2 | 370 | |||
DR003779.1 | - | 2 | 424 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
